Gene Transcripts

Gene Symbol
TXN
Entrez Gene ID
7295
Gene Name
thioredoxin
Gene Aliases
TRDX, TRX, TRX1
Chromosome Location
Chr. 9: 110243812 - 110256640 on Build GRCh38
UniGene ID
Hs.435136
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003329.3 NP_003320.2 3 366
GenBank mRNA AF085844.1 - 3 173
AF276919.1 - 2 161
AF313911.1 - 3 171
AK289508.1 - 3 287
AW160756.1 - 3 366
AY004872.1 - 3 171
BC003377.1 - 3 218 2901084
BC054866.1 - 3 237 6463280
CR407665.1 - 3 161
J04026.1 - 3 224
JQ313905.1 - 3 161
X77584.1 - 3 241

View Full Product Details