Gene Transcripts

Gene Symbol
ETNK1
Entrez Gene ID
55500
Gene Name
ethanolamine kinase 1
Gene Aliases
EKI, EKI 1, EKI1, Nbla10396
Chromosome Location
Chr. 12: 22625142 - 22690674 on Build GRCh38
UniGene ID
Hs.29464
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039481.1 NP_001034570.1 2 513
NM_018638.4 NP_061108.2 2 513
XM_005253420.4 XP_005253477.1 2 510
XM_017019580.1 XP_016875069.1 2 510
XM_017019581.1 XP_016875070.1 2 510
GenBank mRNA AF207600.2 - 2 579
AK074425.1 - 2 458
AL137750.1 - 2 463
BC006111.1 - 2 465
BC066907.1 - 2 513 5260109
BT007084.1 - 2 224

View Full Product Details