Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001005855.1 | NP_001005855.1 | 4 | 350 | |
NM_020452.3 | NP_065185.1 | 4 | 269 | ||
XM_005245355.2 | XP_005245412.1 | 3 | 416 | ||
XM_005245356.3 | XP_005245413.1 | 4 | 369 | ||
XM_017001871.1 | XP_016857360.1 | 3 | 416 | ||
XM_017001872.1 | XP_016857361.1 | 4 | 365 | ||
GenBank mRNA | AK054886.1 | - | 3 | 416 | |
AY302537.1 | - | 4 | 269 | ||
BC069264.1 | - | 4 | 357 |