Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_021194.2 | NP_067017.2 | 2 | 841 | |
GenBank mRNA | AA853540.1 | - | 2 | 171 | |
AB451467.1 | - | 2 | 696 | ||
BC121015.1 | - | 2 | 826 | 40117054 | |
BC121016.1 | - | 2 | 826 | 40117055 | |
BE744293.1 | - | 2 | 388 | ||
CB990012.1 | - | 1 | 53 |