Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001144884.1 | NP_001138356.1 | 10 | 1205 | |
NM_133496.4 | NP_598003.2 | 10 | 1203 | ||
XM_011540779.2 | XP_011539081.1 | 11 | 1302 | ||
XM_017000400.1 | XP_016855889.1 | 10 | 1203 | ||
GenBank mRNA | AF233345.1 | - | 10 | 1033 | |
AF529197.1 | - | 10 | 1018 | ||
AK313638.1 | - | 10 | 1186 | ||
AY094606.1 | - | 10 | 1018 | ||
BC064692.1 | - | 10 | 1159 | 30330665 | |
BX537375.1 | - | 10 | 1205 | ||
CR936806.1 | - | 10 | 1199 |