Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001145290.1 | NP_001138762.1 | 2 | 345 | |
NM_198277.2 | NP_938018.1 | 2 | 345 | ||
GenBank mRNA | AK074100.1 | - | 2 | 127 | |
AK074207.1 | - | 2 | 345 | ||
AK290314.1 | - | 2 | 157 | ||
BC051314.1 | - | 2 | 126 | 6207707 |