Gene Transcripts

Gene Symbol
REV1
Entrez Gene ID
51455
Gene Name
REV1, DNA directed polymerase
Gene Aliases
AIBP80, REV1L
Chromosome Location
Chr. 2: 99400475 - 99490123 on Build GRCh38
UniGene ID
Hs.443077
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001037872.2 NP_001032961.1 4, 5 608
NM_001321454.1 NP_001308383.1 4, 5 608
NM_001321455.1 NP_001308384.1 5, 6 765
NM_001321458.1 NP_001308387.1 4, 5 608
NM_001321459.1 NP_001308388.1 4, 5 608
NM_001321460.1 NP_001308389.1 4, 5 608
NM_016316.3 NP_057400.1 4, 5 608
NR_135649.1 4, 5 608
NR_135650.1 5, 6 765
NR_135651.1 4, 5 608
NR_135652.1 4, 5 608
NR_135653.1 4, 5 608
XM_017004309.1 XP_016859798.1 4, 5 1021
XM_017004310.1 XP_016859799.1 4, 5 387
XM_017004311.1 XP_016859800.1 4, 5 387
XM_017004312.1 XP_016859801.1 4, 5 603
XM_017004313.1 XP_016859802.1 4, 5 2337
GenBank mRNA AB047646.1 - 4, 5 533
AF151538.1 - 4, 5 561
AF206019.1 - 4, 5 570
AF357886.1 - 4, 5 526
AK002087.1 - 2, 3 470
AK307719.1 - 4, 5 519
BC037734.1 - 5, 6 746 4824689
BC130411.1 - 3, 4 379 40146442
DA852908.1 - 1, 2 106
DR000409.1 - 4, 5 499

View Full Product Details