Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001130721.1 | NP_001124193.1 | 5 | 1254 | |
NM_024090.2 | NP_076995.1 | 4 | 1457 | ||
XM_011532233.2 | XP_011530535.1 | 5 | 1260 | ||
XM_011532234.2 | XP_011530536.1 | 5 | 1328 | ||
XM_011532235.2 | XP_011530537.1 | 5 | 1372 | ||
GenBank mRNA | AK027031.1 | - | 4 | 1408 | |
AK074813.1 | - | 4 | 1102 | ||
AK074882.1 | - | 1 | 763 | ||
BC001305.1 | - | 5 | 1241 | 3452423 |