Gene Transcripts

Gene Symbol
DNAI1
Entrez Gene ID
27019
Gene Name
dynein axonemal intermediate chain 1
Gene Aliases
CILD1, DIC1, ICS1, PCD
Chromosome Location
Chr. 9: 34458752 - 34520989 on Build GRCh38
UniGene ID
Hs.112667
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001281428.1 NP_001268357.1 4 451
NM_012144.3 NP_036276.1 4 451
XM_006716758.3 XP_006716821.1 3 149
XM_011517846.1 XP_011516148.1 4 451
XM_011517847.2 XP_011516149.1 4 451
XM_011517848.1 XP_011516150.1 4 451
XM_011517849.1 XP_011516151.1 4 451
XM_011517850.2 XP_011516152.1 4 451
XM_017014625.1 XP_016870114.1 4 451
GenBank mRNA AF091619.1 - 4 368
AK290696.1 - 4 375
AK302499.1 - 4 314
AK303937.1 - 4 340
AK314639.1 - 4 363
BC030583.2 - 4 379 4837991
BM013886.1 - 4 390
DB033871.1 - 4 375
DB046118.1 - 4 452

View Full Product Details