Gene Transcripts
- Gene Symbol
- MYO10
- Entrez Gene ID
- 4651
- Gene Name
- myosin X
- Gene Aliases
- Chromosome Location
- Chr. 5: 16661907 - 16936276 on Build GRCh38
- UniGene ID
- Hs.481720
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_012334.2 |
NP_036466.2
|
17, 18 |
2212 |
|
XM_006714475.2 |
XP_006714538.1
|
16, 17 |
1919 |
|
GenBank mRNA |
AB018342.2 |
- |
17, 18 |
2199 |
|
AF132021.1 |
- |
17, 18 |
1824 |
|
AF234532.1 |
- |
17, 18 |
1966 |
|
AF247457.2 |
- |
17, 18 |
1897 |
|
AK024719.1 |
- |
8, 9 |
823 |
|
AK125246.1 |
- |
17, 18 |
2196 |
|
AL832428.1 |
- |
7, 8 |
669 |
|
BC014956.1 |
- |
17, 18 |
2208 |
|
BC137168.1 |
- |
17, 18 |
1921 |
9021165 |
BC150285.1 |
- |
17, 18 |
2199 |
8860110 |
View Full Product Details