Gene Transcripts

Gene Symbol
MYO5C
Entrez Gene ID
55930
Gene Name
myosin VC
Gene Aliases
Chromosome Location
Chr. 15: 52192318 - 52295798 on Build GRCh38
UniGene ID
Hs.487036
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018728.3 NP_061198.2 6 873
XM_011521781.2 XP_011520083.1 6 873
XM_017022408.1 XP_016877897.1 6 873
XM_017022409.1 XP_016877898.1 2 262
XM_017022410.1 XP_016877899.1 6 845
GenBank mRNA AF272390.1 - 6 730
AK301079.1 - 5 733
AK304136.1 - 7 1028
BC064841.1 - 6 819
BC144611.1 - 6 745

View Full Product Details