Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_004521.2 | NP_004512.1 | 2 | 642 | |
XM_017016224.1 | XP_016871713.1 | 2 | 248 | ||
GenBank mRNA | AK289413.1 | - | 2 | 588 | |
AK307502.1 | - | 3 | 339 | ||
BC010885.1 | - | 2 | 578 | ||
BC065267.1 | - | 2 | 599 | ||
BC080604.1 | - | 2 | 573 | ||
BC111042.1 | - | 2 | 582 | ||
BC126279.1 | - | 2 | 395 | 8991995 | |
BC126281.1 | - | 2 | 395 | 8991997 | |
FJ224324.1 | - | 2 | 485 | ||
X65873.1 | - | 2 | 485 |