Gene Transcripts

Gene Symbol
KIF2B
Entrez Gene ID
84643
Gene Name
kinesin family member 2B
Gene Aliases
Chromosome Location
Chr. 17: 53822878 - 53825212 on Build GRCh38
UniGene ID
Hs.226805
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032559.4 NP_115948.4 1 304
GenBank mRNA AF333335.2 - 1 282
BC033802.1 - 1 242 5170969
DB457485.1 - 1 304

View Full Product Details