Gene Transcripts

Gene Symbol
SERPINA7
Entrez Gene ID
6906
Gene Name
serpin family A member 7
Gene Aliases
TBG
Chromosome Location
Chr. X: 106032439 - 106038727 on Build GRCh38
UniGene ID
Hs.76838
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000354.5 NP_000345.2 5 1201
XM_006724683.2 XP_006724746.1 5 1229
GenBank mRNA BC020747.2 - 4 1170
M14091.1 - 4 1484

View Full Product Details