Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001300960.1 | NP_001287889.1 | 2 | 360 | |
NM_001300963.1 | NP_001287892.1 | 3 | 176 | ||
NM_001300964.1 | NP_001287893.1 | 2 | 130 | ||
NM_015076.4 | NP_055891.1 | 2 | 360 | ||
XM_011535630.2 | XP_011533932.1 | 3 | 838 | ||
XM_011535631.2 | XP_011533933.1 | 3 | 830 | ||
XM_011535632.1 | XP_011533934.1 | 1 | 52 | ||
XM_017010587.1 | XP_016866076.1 | 2 | 305 | ||
GenBank mRNA | AB028951.2 | - | 2 | 177 | |
AK292360.1 | - | 2 | 268 | ||
AY028424.1 | - | 2 | 360 | ||
BC024247.1 | - | 3 | 181 | ||
BC037289.1 | - | 2 | 255 | 5266531 | |
JX028238.1 | - | 2 | 180 |