Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001195643.1 | NP_001182572.1 | 2 | 1779 | |
NM_176815.4 | NP_789785.1 | 2 | 1666 | ||
XM_011512537.2 | XP_011510839.1 | 2 | 1844 | ||
GenBank mRNA | AK122674.1 | - | 2 | 1779 | |
AL832912.1 | - | 2 | 1455 | ||
BC045541.1 | - | 2 | 1497 | 5262102 | |
BC063379.1 | - | 2 | 1455 | 4109721 |