Gene Transcripts

Gene Symbol
ARSE
Entrez Gene ID
415
Gene Name
arylsulfatase E (chondrodysplasia punctata 1)
Gene Aliases
ASE, CDPX, CDPX1, CDPXR
Chromosome Location
Chr. X: 2934632 - 2968310 on Build GRCh38
UniGene ID
Hs.386975
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000047.2 NP_000038.2 8 1295
NM_001282628.1 NP_001269557.1 9 1581
NM_001282631.1 NP_001269560.1 7 1065
XM_005274518.2 XP_005274575.1 8 1222
XM_005274519.4 XP_005274576.1 8 1385
XM_005274521.4 XP_005274578.1 7 1178
XM_011545521.1 XP_011543823.1 6 865
XM_017029525.1 XP_016885014.1 9 1512
XM_017029526.1 XP_016885015.1 7 1065
GenBank mRNA AK223183.1 - 8 1125
AK223199.1 - 8 1092
AK293648.1 - 8 1000
AK301004.1 - 9 1412
AK313093.1 - 8 1114
BC130438.1 - 8 1090 40146469
X83573.1 - 8 1112

View Full Product Details