Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_005908.3 | NP_005899.3 | 4 | 627 | |
XM_017008203.1 | XP_016863692.1 | 5 | 613 | ||
XM_017008204.1 | XP_016863693.1 | 4 | 582 | ||
GenBank mRNA | AB209846.1 | - | 4 | 574 | |
AK291598.1 | - | 4 | 615 | ||
AK312827.1 | - | 4 | 582 | ||
AU132611.1 | - | 4 | 627 | ||
BC015743.1 | - | 4 | 617 | 4905535 | |
EU009130.1 | - | 4 | 527 | ||
U60337.1 | - | 4 | 614 |