Gene Transcripts

Gene Symbol
EYA2
Entrez Gene ID
2139
Gene Name
EYA transcriptional coactivator and phosphatase 2
Gene Aliases
EAB1
Chromosome Location
Chr. 20: 46894624 - 47188844 on Build GRCh38
UniGene ID
Hs.472877
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005244.4 NP_005235.3 15 1839
NM_172110.3 NP_742108.2 13 1602
XM_005260327.2 XP_005260384.1 13 1286
XM_017027721.1 XP_016883210.1 14 1467
GenBank mRNA AF055015.1 - 11 1153
AJ007992.1 - 4 334
AK093582.1 - 6 1723
AK293032.1 - 15 1543
AY705349.1 - 13 1385
BC000289.2 - 9 991
BC008803.2 - 15 1521 3944547
BC013882.2 - 13 1602
BT006682.1 - 14 1393
U71207.1 - 14 1467
U81601.1 - 6 583

View Full Product Details