Gene Transcripts

Gene Symbol
IDUA
Entrez Gene ID
3425
Gene Name
iduronidase, alpha-L-
Gene Aliases
IDA, MPS1
Chromosome Location
Chr. 4: 986997 - 1004557 on Build GRCh38
UniGene ID
Hs.89560
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000203.4 NP_000194.2 3 419
NR_110313.1 3 419
XM_011513461.2 XP_011511763.1 2 564
XM_017008163.1 XP_016863652.1 3 367
GenBank mRNA AK098805.1 - 2 387
AK125223.1 - 2 231
AK291816.1 - 3 352
BC029959.1 - 3 343
DA057148.1 - 3 367
M74715.1 - 3 419

View Full Product Details