Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001040427.1 | NP_001035517.1 | 10 | 1533 | |
NM_001286249.1 | NP_001273178.1 | 8 | 1328 | ||
NM_005326.4 | NP_005317.2 | 9 | 1437 | ||
XM_011522469.2 | XP_011520771.1 | 9 | 1141 | ||
XM_011522470.2 | XP_011520772.1 | 8 | 1030 | ||
XM_017023195.1 | XP_016878684.1 | 9 | 1066 | ||
GenBank mRNA | AK054943.1 | - | 3 | 1400 | |
AK298174.1 | - | 8 | 968 | ||
AK299173.1 | - | 9 | 1068 | ||
AK299993.1 | - | 9 | 1068 | ||
BC000840.1 | - | 9 | 1057 | 3457214 | |
BC002627.2 | - | 9 | 1386 | 3608098 | |
X90999.1 | - | 8 | 922 |