Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001282225.1 | NP_001269154.1 | 5 | 1061 | |
NM_001282226.1 | NP_001269155.1 | 5 | 1028 | ||
NM_001282227.1 | NP_001269156.1 | 5 | 834 | ||
NM_001282228.1 | NP_001269157.1 | 5 | 936 | ||
NM_001282229.1 | NP_001269158.1 | 4 | 693 | ||
NM_177405.2 | NP_803124.1 | 2 | 267 | ||
XM_006724080.3 | XP_006724143.1 | 5 | 648 | ||
XM_011546133.1 | XP_011544435.1 | 5 | 953 | ||
GenBank mRNA | AB209648.1 | - | 5 | 648 | |
AF190746.1 | - | 4 | 997 | ||
AK027682.1 | - | 2 | 88 | ||
AK074702.1 | - | 2 | 139 | ||
AK292689.1 | - | 5 | 1049 | ||
AK295174.1 | - | 5 | 785 | ||
AK304818.1 | - | 5 | 828 | ||
AK314321.1 | - | 5 | 852 | ||
BC051755.1 | - | 4 | 832 | 5753141 | |
CR456417.1 | - | 4 | 819 | ||
DA864284.1 | - | 2 | 134 | ||
DB130953.1 | - | 2 | 267 |