Gene Transcripts

Gene Symbol
SLC9A2
Entrez Gene ID
6549
Gene Name
solute carrier family 9 member A2
Gene Aliases
NHE2
Chromosome Location
Chr. 2: 102619689 - 102711350 on Build GRCh38
UniGene ID
Hs.250083
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AF073299.1 - 2 733
BC136377.1 - 2 803 9020364
BC136378.1 - 2 803 9020365

View Full Product Details