Gene Transcripts
- Gene Symbol
- PIGA
- Entrez Gene ID
- 5277
- Gene Name
- phosphatidylinositol glycan anchor biosynthesis class A
- Gene Aliases
- GPI3, MCAHS2, PIG-A, PNH1
- Chromosome Location
- Chr. X: 15319451 - 15335554 on Build GRCh38
- UniGene ID
- Hs.137154
- Species
- Homo sapiens
- Gene Symbol
- PIGAP1
- Entrez Gene ID
- 5278
- Gene Name
- phosphatidylinositol glycan anchor biosynthesis class A pseudogene 1
- Gene Aliases
- PIG-A-II
- Chromosome Location
- Chr. 12: 100578634 - 100582672 on Build GRCh38
- UniGene ID
- Hs.20575
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
S78467.1 |
- |
1 |
865 |
|
View Full Product Details