Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_005074.3 | NP_005065.2 | 7 | 805 | |
XM_011514818.2 | XP_011513120.2 | 7 | 842 | ||
XM_011514819.2 | XP_011513121.2 | 6 | 755 | ||
XM_011514820.2 | XP_011513122.2 | 7 | 842 | ||
XM_011514821.2 | XP_011513123.1 | 5 | 559 | ||
XM_017011199.1 | XP_016866688.1 | 7 | 842 | ||
XM_017011200.1 | XP_016866689.1 | 7 | 842 | ||
XM_017011201.1 | XP_016866690.1 | 7 | 842 | ||
XM_017011202.1 | XP_016866691.1 | 6 | 770 | ||
GenBank mRNA | AK290783.1 | - | 7 | 787 | |
AK308035.1 | - | 7 | 787 | ||
AY780791.1 | - | 6 | 689 | ||
BC101745.1 | - | 6 | 742 | 8069251 | |
BC101747.1 | - | 6 | 742 | 8069253 | |
X71355.1 | - | 6 | 701 |