Gene Transcripts

Gene Symbol
SLCO6A1
Entrez Gene ID
133482
Gene Name
solute carrier organic anion transporter family member 6A1
Gene Aliases
CT48, GST, OATP-I, OATP6A1, OATPY
Chromosome Location
Chr. 5: 102371782 - 102502213 on Build GRCh38
UniGene ID
Hs.388874
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001289002.1 NP_001275931.1 2 602
NM_001289004.1 NP_001275933.1 2 602
NM_001308014.1 NP_001294943.1 2 602
NM_173488.4 NP_775759.3 2 602
XM_005271874.3 XP_005271931.1 2 3799
XM_011543147.2 XP_011541449.1 2 3799
XM_011543148.2 XP_011541450.1 2 3799
XM_011543153.2 XP_011541455.1 1 23
XM_011543154.2 XP_011541456.1 2 3799
GenBank mRNA AF505657.1 - 2 573
AK131445.1 - 2 499
AY273897.1 - 2 565
BC034976.2 - 2 604 4820512
BG772379.1 - 2 592
BM554439.1 - 2 589

View Full Product Details