Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001143838.2 | NP_001137310.1 | 11 | 1914 | |
NM_001284509.1 | NP_001271438.1 | 12 | 2001 | ||
NM_001284510.1 | NP_001271439.1 | 11 | 1923 | ||
NM_177550.4 | NP_808218.1 | 12 | 2052 | ||
GenBank mRNA | AK124317.1 | - | 1 | 3407 | |
AK127797.1 | - | 12 | 1949 | ||
AK172779.1 | - | 5 | 905 | ||
AK172785.1 | - | 12 | 1997 | ||
AY151833.1 | - | 12 | 1976 | ||
BX647354.1 | - | 1 | 4157 | ||
CR749584.1 | - | 7 | 1166 |