Gene Transcripts

Gene Symbol
RAB1A
Entrez Gene ID
5861
Gene Name
RAB1A, member RAS oncogene family
Gene Aliases
RAB1, YPT1
Chromosome Location
Chr. 2: 65086854 - 65130301 on Build GRCh38
UniGene ID
Hs.310645
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004161.4 NP_004152.1 6 1736
NM_015543.1 NP_056358.1 4 1508
XM_005264468.1 XP_005264525.1 5 1445
GenBank mRNA AK055927.1 - 4 1351

View Full Product Details