Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_002806.3 | NP_002797.3 | 5 | 333 | |
XM_017021472.1 | XP_016876961.1 | 5 | 1836 | ||
GenBank mRNA | AF006305.1 | - | 5 | 306 | |
AK299155.1 | - | 4 | 247 | ||
AK313670.1 | - | 5 | 301 | ||
BC005390.1 | - | 5 | 311 | 3996906 | |
BM788029.1 | - | 5 | 333 | ||
BT006843.1 | - | 5 | 291 | ||
CR456709.1 | - | 5 | 291 | ||
D78275.1 | - | 5 | 307 | ||
FJ224299.1 | - | 5 | 307 |