Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_015983.3 | NP_057067.1 | 7 | 1369 | |
XM_006715734.3 | XP_006715797.1 | 8 | 956 | ||
XM_011515420.2 | XP_011513722.1 | 8 | 1462 | ||
XM_011515422.2 | XP_011513724.1 | 6 | 1306 | ||
XM_011515423.2 | XP_011513725.1 | 5 | 740 | ||
XM_017012292.1 | XP_016867781.1 | 9 | 1049 | ||
XM_017012295.1 | XP_016867784.1 | 6 | 803 | ||
GenBank mRNA | AF125044.1 | - | 7 | 1408 | |
AF125045.1 | - | 6 | 1255 | ||
AK001446.1 | - | 7 | 1357 | ||
BC004104.2 | - | 7 | 1361 | 3835609 |