Gene Transcripts
- Gene Symbol
- ATP9B
- Entrez Gene ID
- 374868
- Gene Name
- ATPase phospholipid transporting 9B (putative)
- Gene Aliases
- ATPASEP, ATPIIB, HUSSY-20, NEO1L, hMMR1
- Chromosome Location
- Chr. 18: 79069275 - 79378391 on Build GRCh38
- UniGene ID
- Hs.465475
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001306085.1 |
NP_001293014.1
|
7, 8 |
919 |
|
NM_198531.4 |
NP_940933.3
|
7, 8 |
919 |
|
XM_011525963.2 |
XP_011524265.1
|
8, 9 |
1085 |
|
XM_011525964.2 |
XP_011524266.1
|
7, 8 |
800 |
|
XM_011525965.2 |
XP_011524267.1
|
8, 9 |
1081 |
|
XM_011525966.2 |
XP_011524268.1
|
7, 8 |
1036 |
|
XM_011525972.2 |
XP_011524274.1
|
2, 3 |
160 |
|
XM_017025726.1 |
XP_016881215.1
|
8, 9 |
1085 |
|
XM_017025727.1 |
XP_016881216.1
|
8, 9 |
1085 |
|
XM_017025728.1 |
XP_016881217.1
|
8, 9 |
1085 |
|
XM_017025729.1 |
XP_016881218.1
|
8, 9 |
1085 |
|
XM_017025730.1 |
XP_016881219.1
|
8, 9 |
1085 |
|
XM_017025731.1 |
XP_016881220.1
|
8, 9 |
1085 |
|
XM_017025732.1 |
XP_016881221.1
|
8, 9 |
1085 |
|
XM_017025733.1 |
XP_016881222.1
|
8, 9 |
1085 |
|
XM_017025734.1 |
XP_016881223.1
|
8, 9 |
1085 |
|
XM_017025735.1 |
XP_016881224.1
|
8, 9 |
1085 |
|
XM_017025736.1 |
XP_016881225.1
|
8, 9 |
1085 |
|
XM_017025737.1 |
XP_016881226.1
|
8, 9 |
1085 |
|
XM_017025739.1 |
XP_016881228.1
|
5, 6 |
490 |
|
XM_017025742.1 |
XP_016881231.1
|
8, 9 |
1085 |
|
GenBank mRNA |
AB373110.1 |
- |
6, 7 |
627 |
|
AK308291.1 |
- |
7, 8 |
801 |
|
BC125219.1 |
- |
7, 8 |
797 |
40123958 |
BC125220.1 |
- |
7, 8 |
797 |
40123959 |
View Full Product Details