Gene Transcripts

Gene Symbol
RAB40A
Entrez Gene ID
142684
Gene Name
RAB40A, member RAS oncogene family
Gene Aliases
RAR2, RAR2A
Chromosome Location
Chr. X: 103493266 - 103519489 on Build GRCh38
UniGene ID
Hs.706904
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_080879.2 NP_543155.2 3 1286
XM_005262083.4 XP_005262140.1 2 1763
XM_011530872.2 XP_011529174.1 2 1760
GenBank mRNA AF132748.1 - 1 1009
AF422143.1 - 3 1289
AK056859.1 - 1 3063

View Full Product Details