Gene Transcripts

Gene Symbol
RAB37
Entrez Gene ID
326624
Gene Name
RAB37, member RAS oncogene family
Gene Aliases
Chromosome Location
Chr. 17: 74671117 - 74747335 on Build GRCh38
UniGene ID
Hs.351413
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001006638.2 NP_001006639.1 7 516
NM_001163989.1 NP_001157461.1 7 665
NM_001163990.1 NP_001157462.1 6 405
NM_175738.4 NP_783865.1 7 909
XM_005257288.4 XP_005257345.1 7 534
GenBank mRNA AK054846.1 - 6 1345
AK057069.1 - 1 60
AK098068.1 - 7 516
AK127073.1 - 7 1448
AK290202.1 - 7 885
AK296062.1 - 7 504
AK296172.1 - 6 368
AK298033.1 - 5 900
AK303442.1 - 7 568
BC016615.1 - 7 462 4520191
BC040547.1 - 7 895 5287038

View Full Product Details