Gene Transcripts
- Gene Symbol
- IFT27
- Entrez Gene ID
- 11020
- Gene Name
- intraflagellar transport 27
- Gene Aliases
- BBS19, RABL4, RAYL
- Chromosome Location
- Chr. 22: 36758202 - 36776133 on Build GRCh38
- UniGene ID
- Hs.415172
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001177701.2 |
NP_001171172.1
|
2, 3 |
538 |
|
NM_006860.4 |
NP_006851.1
|
2, 3 |
535 |
|
XM_006724106.2 |
XP_006724169.1
|
3, 4 |
299 |
|
XM_017028540.1 |
XP_016884029.1
|
2, 3 |
542 |
|
GenBank mRNA |
AB209424.1 |
- |
2, 3 |
530 |
|
AK307105.1 |
- |
2, 3 |
498 |
|
AK307382.1 |
- |
2, 3 |
507 |
|
AL022729.1 |
- |
2, 3 |
479 |
|
BC000566.2 |
- |
2, 3 |
516 |
3163341 |
BM711121.1 |
- |
2, 3 |
126 |
|
BT006815.1 |
- |
2, 3 |
112 |
|
CR456558.1 |
- |
2, 3 |
154 |
|
View Full Product Details