Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_020798.2 | NP_065849.1 | 4 | 1276 | |
XM_011545187.2 | XP_011543489.1 | 4 | 419 | ||
XM_011545188.2 | XP_011543490.1 | 2 | 202 | ||
XM_017018052.1 | XP_016873541.1 | 4 | 1273 | ||
XM_017018053.1 | XP_016873542.1 | 3 | 268 | ||
GenBank mRNA | AB037793.1 | - | 1 | 739 | |
AJ586137.1 | - | 3 | 917 | ||
AK307875.1 | - | 3 | 502 | ||
DB095282.1 | - | 3 | 532 |