Gene Transcripts

Gene Symbol
ANXA3
Entrez Gene ID
306
Gene Name
annexin A3
Gene Aliases
ANX3
Chromosome Location
Chr. 4: 78551588 - 78610451 on Build GRCh38
UniGene ID
Hs.480042
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005139.2 NP_005130.1 12, 13 1222
GenBank mRNA AB209868.1 - 1, 2 2536
AK313945.1 - 12, 13 997
BC000871.1 - 12, 13 1054 3460409
CR407648.1 - 10, 11 912
M20560.1 - 11, 12 958
M63310.1 - 11, 12 948

View Full Product Details