Gene Transcripts
- Gene Symbol
- SFXN1
- Entrez Gene ID
- 94081
- Gene Name
- sideroflexin 1
- Gene Aliases
- TCC
- Chromosome Location
- Chr. 5: 175478498 - 175529742 on Build GRCh38
- UniGene ID
- Hs.369440
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001322977.1 |
NP_001309906.1
|
5, 6 |
697 |
|
NM_001322978.1 |
NP_001309907.1
|
5, 6 |
640 |
|
NM_001322980.1 |
NP_001309909.1
|
5, 6 |
669 |
|
NM_001322981.1 |
NP_001309910.1
|
5, 6 |
697 |
|
NM_001322982.1 |
NP_001309911.1
|
5, 6 |
668 |
|
NM_001322983.1 |
NP_001309912.1
|
5, 6 |
669 |
|
NM_022754.6 |
NP_073591.2
|
5, 6 |
669 |
|
GenBank mRNA |
AF327346.1 |
- |
5, 6 |
579 |
|
AK022287.1 |
- |
5, 6 |
731 |
|
AK022938.1 |
- |
3, 4 |
303 |
|
AK056915.1 |
- |
5, 6 |
574 |
|
BC020517.1 |
- |
4, 5 |
464 |
3888861 |
BC063241.1 |
- |
5, 6 |
587 |
6187994 |
BX648188.1 |
- |
5, 6 |
585 |
|
View Full Product Details