Gene Transcripts

Gene Symbol
TSPAN15
Entrez Gene ID
23555
Gene Name
tetraspanin 15
Gene Aliases
2700063A19Rik, NET-7, NET7, TM4SF15
Chromosome Location
Chr. 10: 69451464 - 69549508 on Build GRCh38
UniGene ID
Hs.499941
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012339.3 NP_036471.1 2 277
XM_017016010.1 XP_016871499.1 2 283
GenBank mRNA AF120266.1 - 2 268
BC003157.1 - 2 271 2958221
BC004161.2 - 2 271 2958221

View Full Product Details