Gene Transcripts

Gene Symbol
SPINK4
Entrez Gene ID
27290
Gene Name
serine peptidase inhibitor, Kazal type 4
Gene Aliases
HEL136, PEC-60, PEC60
Chromosome Location
Chr. 9: 33240198 - 33248567 on Build GRCh38
UniGene ID
Hs.555934
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014471.1 NP_055286.1 2 103
GenBank mRNA AF048700.1 - 2 103
BC110068.1 - 2 103 40013921
EU794680.1 - 2 103

View Full Product Details