Gene Transcripts

Gene Symbol
OSBPL3
Entrez Gene ID
26031
Gene Name
oxysterol binding protein like 3
Gene Aliases
ORP-3, ORP3, OSBP3
Chromosome Location
Chr. 7: 24796537 - 24980218 on Build GRCh38
UniGene ID
Hs.520259
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015550.3 NP_056365.1 4, 5 741
NM_145320.2 NP_663160.1 3, 4 414
NM_145321.2 NP_663161.1 3, 4 414
NM_145322.2 NP_663162.1 3, 4 414
NR_104111.1 3, 4 414
NR_104112.1 3, 4 414
XM_005249698.3 XP_005249755.1 4, 5 461
XM_006715681.3 XP_006715744.1 4, 5 728
XM_006715682.3 XP_006715745.1 4, 5 747
XM_006715683.3 XP_006715746.1 4, 5 747
XM_006715684.3 XP_006715747.1 3, 4 414
XM_011515258.2 XP_011513560.1 4, 5 538
XM_011515259.2 XP_011513561.1 5, 6 1035
XM_017011948.1 XP_016867437.1 6, 7 898
XM_017011949.1 XP_016867438.1 5, 6 1035
GenBank mRNA AB014604.2 - 4, 5 678
AF323727.1 - 4, 5 536
AF392444.1 - 4, 5 650
AF491781.1 - 3, 4 368
AF491782.1 - 3, 4 368
AF491783.1 - 3, 4 368
AF491784.1 - 3, 4 368
AF491785.1 - 3, 4 368
AF491786.1 - 3, 4 368
AL832293.1 - 2, 3 83
AY008372.1 - 4, 5 571
BC017731.1 - 4, 5 670 3909164

View Full Product Details