Gene Transcripts

Gene Symbol
ANGPTL7
Entrez Gene ID
10218
Gene Name
angiopoietin like 7
Gene Aliases
AngX, CDT6, dJ647M16.1
Chromosome Location
Chr. 1: 11189289 - 11195981 on Build GRCh38
UniGene ID
Hs.146559
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021146.3 NP_066969.1 5 1526
GenBank mRNA AB593026.1 - 5 1460
AB593027.1 - 5 1527
AY358301.1 - 5 1449
BC001881.1 - 5 1474 3544149
Y16132.1 - 5 1242

View Full Product Details