Gene Transcripts
- Gene Symbol
- CDC25B
- Entrez Gene ID
- 994
- Gene Name
- cell division cycle 25B
- Gene Aliases
- Chromosome Location
- Chr. 20: 3786772 - 3806121 on Build GRCh38
- UniGene ID
- Hs.153752
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001287516.1 |
NP_001274445.1
|
4, 5 |
583 |
|
NM_001287517.1 |
NP_001274446.1
|
4, 5 |
541 |
|
NM_001287518.1 |
NP_001274447.1
|
4, 5 |
583 |
|
NM_001287519.1 |
NP_001274448.1
|
4, 5 |
314 |
|
NM_001287520.1 |
NP_001274449.1
|
4, 5 |
356 |
|
NM_001287522.1 |
NP_001274451.1
|
4, 5 |
356 |
|
NM_001287524.1 |
NP_001274453.1
|
1, 2 |
261 |
|
NM_004358.4 |
NP_004349.1
|
4, 5 |
1166 |
|
NM_021872.3 |
NP_068658.1
|
4, 5 |
1208 |
|
NM_021873.3 |
NP_068659.1
|
4, 5 |
1208 |
|
NR_136335.1 |
|
4, 5 |
356 |
|
NR_136336.1 |
|
4, 5 |
583 |
|
GenBank mRNA |
AK092713.1 |
- |
4, 5 |
356 |
|
AK295573.1 |
- |
4, 5 |
425 |
|
AK299028.1 |
- |
4, 5 |
390 |
|
AK299192.1 |
- |
4, 5 |
359 |
|
BC006395.2 |
- |
4, 5 |
489 |
4135465 |
BC009953.2 |
- |
4, 5 |
569 |
4130804 |
BC051711.1 |
- |
4, 5 |
1113 |
5301205 |
BX640836.1 |
- |
1, 2 |
253 |
|
BX647988.1 |
- |
4, 5 |
271 |
|
DA065196.1 |
- |
1, 2 |
261 |
|
FR695900.1 |
- |
4, 5 |
356 |
|
FR695901.1 |
- |
4, 5 |
356 |
|
M81934.1 |
- |
4, 5 |
445 |
|
S78187.1 |
- |
4, 5 |
613 |
|
Z68092.1 |
- |
4, 5 |
516 |
|
View Full Product Details