Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_031300.3 | NP_112590.1 | 6 | 1187 | |
GenBank mRNA | AF114834.1 | - | 6 | 782 | |
AK092842.1 | - | 1 | 581 | ||
AK222579.1 | - | 6 | 792 | ||
AK303397.1 | - | 5 | 871 | ||
AK316468.1 | - | 5 | 868 | ||
AL833959.1 | - | 3 | 1833 | ||
BC000745.2 | - | 6 | 776 | 2821596 | |
BC032586.1 | - | 5 | 868 |