Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001289000.1 | NP_001275929.1 | 10 | 1219 | |
NM_001289001.1 | NP_001275930.1 | 10 | 1229 | ||
NM_153609.3 | NP_705837.1 | 10 | 1298 | ||
XM_011529989.2 | XP_011528291.1 | 7 | 925 | ||
GenBank mRNA | AJ319876.1 | - | 9 | 1161 | |
AY055384.1 | - | 10 | 1298 | ||
AY358398.1 | - | 10 | 1229 | ||
BC039082.1 | - | 10 | 1301 | 5164881 | |
CR456446.1 | - | 10 | 1219 |