Gene Transcripts

Gene Symbol
ZC3H11A
Entrez Gene ID
9877
Gene Name
zinc finger CCCH-type containing 11A
Gene Aliases
ZC3HDC11A
Chromosome Location
Chr. 1: 203795537 - 203854128 on Build GRCh38
UniGene ID
Hs.532399
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001319238.1 NP_001306167.1 15 2420
NM_001319239.1 NP_001306168.1 15 2315
NM_014827.5 NP_055642.3 15 1991
XM_005245643.2 XP_005245700.1 15 2010
XM_005245644.3 XP_005245701.1 14 1694
XM_006711673.2 XP_006711736.1 15 1857
XM_011510199.1 XP_011508501.1 13 3062
XM_011510200.1 XP_011508502.1 14 2082
XM_011510201.1 XP_011508503.1 15 2268
XM_011510202.1 XP_011508504.1 13 2007
XM_017002956.1 XP_016858445.1 14 3316
XM_017002957.1 XP_016858446.1 14 3168
XM_017002958.1 XP_016858447.1 14 3155
XM_017002959.1 XP_016858448.1 15 2228
XM_017002960.1 XP_016858449.1 14 3316
XM_017002961.1 XP_016858450.1 13 3062
XM_017002962.1 XP_016858451.1 14 2082
XM_017002963.1 XP_016858452.1 15 1874
XM_017002964.1 XP_016858453.1 14 3228
XM_017002965.1 XP_016858454.1 13 2557
GenBank mRNA AB014563.1 - 12 1502
AK095858.1 - 15 2289
AK296983.1 - 11 1349
AK310470.1 - 3 384
BC014268.2 - 15 1829 3844154
BC038513.2 - 15 2146
BX649148.1 - 12 5607
CR627439.1 - 14 3012
CR627446.1 - 12 5115

Gene Symbol
LOC441155
Entrez Gene ID
441155
Gene Name
zinc finger CCCH-type domain-containing-like
Gene Aliases
Chromosome Location
Chr. 6: 65301418 - 65306069 on Build GRCh38
UniGene ID
Hs.546686
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271675.1 NP_001258604.1 2 2023
XM_017010889.1 XP_016866378.1 2 1539
GenBank mRNA AK302514.1 - 2 1979

View Full Product Details