Gene Transcripts

Gene Symbol
SPEN
Entrez Gene ID
23013
Gene Name
spen family transcriptional repressor
Gene Aliases
HIAA0929, MINT, RBM15C, SHARP
Chromosome Location
Chr. 1: 15847864 - 15940455 on Build GRCh38
UniGene ID
Hs.744843
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015001.2 NP_055816.2 2 396
GenBank mRNA AF356524.1 - 2 396

View Full Product Details