Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001042549.1 | NP_001036014.1 | 7 | 1200 | |
NM_001297736.1 | NP_001284665.1 | 4 | 992 | ||
NM_001297737.1 | NP_001284666.1 | 5 | 1047 | ||
NM_015471.3 | NP_056286.3 | 6 | 1115 | ||
XM_017000968.1 | XP_016856457.1 | 7 | 2611 | ||
XM_017000969.1 | XP_016856458.1 | 7 | 2737 | ||
GenBank mRNA | AF201941.1 | - | 6 | 1087 | |
AF255793.1 | - | 6 | 1101 | ||
AK222996.1 | - | 6 | 1085 | ||
AK223260.1 | - | 6 | 1095 | ||
AK303250.1 | - | 4 | 972 | ||
AL050084.1 | - | 6 | 894 | ||
BC007067.1 | - | 6 | 1089 | 3997280 | |
BC036048.1 | - | 6 | 1093 |