Gene Transcripts

Gene Symbol
HERC4
Entrez Gene ID
26091
Gene Name
HECT and RLD domain containing E3 ubiquitin protein ligase 4
Gene Aliases
Chromosome Location
Chr. 10: 67921899 - 68075348 on Build GRCh38
UniGene ID
Hs.51891
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001278185.1 NP_001265114.1 6 914
NM_001278186.1 NP_001265115.1 5 754
NM_015601.3 NP_056416.2 6 914
NM_022079.2 NP_071362.1 6 914
XM_011539592.2 XP_011537894.1 7 988
XM_011539593.2 XP_011537895.1 7 988
XM_011539594.2 XP_011537896.1 8 1074
XM_011539595.2 XP_011537897.1 8 1124
XM_011539596.2 XP_011537898.1 7 1002
XM_011539597.2 XP_011537899.1 7 988
XM_017016041.1 XP_016871530.1 6 916
GenBank mRNA AB046813.1 - 3 768
AK292809.1 - 6 794
AY221963.1 - 6 851
AY650032.1 - 5 691
AY650033.1 - 6 851
BC015043.1 - 5 657
BC039600.1 - 6 824 5212048
BX537563.1 - 4 592

View Full Product Details