Gene Transcripts

Gene Symbol
TMEM69
Entrez Gene ID
51249
Gene Name
transmembrane protein 69
Gene Aliases
C1orf154
Chromosome Location
Chr. 1: 45688175 - 45694436 on Build GRCh38
UniGene ID
Hs.699548
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016486.3 NP_057570.2 3 436
GenBank mRNA AF151063.1 - 3 415
AK024682.1 - 3 430
BC013608.1 - 3 418 3864518
BC034147.1 - 3 416 4655713
BC039718.2 - 2 339 5784175
BC046130.1 - 3 399 5787422
BC054041.1 - 1 150
BC064539.1 - 3 419 5725426
BC073836.1 - 2 336 5548608
BC121117.2 - 3 378 40120483
BC133692.1 - 3 377 40120480

View Full Product Details