Gene Transcripts

Gene Symbol
C1orf27
Entrez Gene ID
54953
Gene Name
chromosome 1 open reading frame 27
Gene Aliases
ODR4, TTG1, odr-4
Chromosome Location
Chr. 1: 186375758 - 186421375 on Build GRCh38
UniGene ID
Hs.371210
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164245.1 NP_001157717.1 5 647
NM_001164246.1 NP_001157718.1 4 551
NM_017847.5 NP_060317.3 5 647
XM_011509660.1 XP_011507962.1 5 646
XM_011509661.1 XP_011507963.1 5 646
XM_017001543.1 XP_016857032.1 5 646
GenBank mRNA AK000512.1 - 5 567
AK026086.1 - 3 217
AK298107.1 - 4 487
AY854248.1 - 5 540
BC003397.1 - 5 480 3448518
DQ323046.1 - 4 411
DQ529299.1 - 4 411

View Full Product Details